2,4-Dibromo-6-fluorobenzyl bromide ≥95%

Supplier: APOLLO SCIENTIFIC

PC0746-1G PC0746-250MG
76622-264EA 536.84 USD
76622-264 76622-266
2,4-Dibromo-6-fluorobenzyl bromide ≥95%
2,4-Dibromo-6-fluorobenzyl bromide
Formula: C₇H₄Br₃F
MDL Number: MFCD03094091
CAS Number: 497181-23-4
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR