3-Chloro-5-iodophenacyl bromide 95%

Supplier: APOLLO SCIENTIFIC

OR400227-1G OR400227-250MG OR400227-5G
76621-176EA 604.4 USD
76621-176 76621-178 77409-454
3-Chloro-5-iodophenacyl bromide 95%
3-Chloro-5-iodophenacyl bromide
Formula: C₈H₅BrClIO
MDL Number: MFCD28040499
CAS Number: 1823925-41-2
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR