2-Bromo-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: APOLLO SCIENTIFIC

PC912791-1G PC912791-5G
76610-138EA 55.02 USD
76610-138 76610-140
2-Bromo-3-(trifluoromethyl)benzoic acid ≥95%
2-Bromo-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
Melting Pt: 143…145 °C
MDL Number: MFCD08532493
CAS Number: 177420-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR