Palladium on carbon 10% Pd, powder

Supplier: APOLLO SCIENTIFIC
Danger

IN2843-25G IN2843-5G
76610-794EA 412.56 USD
76610-794 76610-796
Palladium on carbon 10% Pd, powder
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 1325
ADR: 4,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR