Iron(II) chloride tetrahydrate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

IN2059-100G IN2059-250G
76615-084EA 55.02 USD
76615-084 76615-086 76615-088
Iron(II) chloride tetrahydrate
Iron (II) chloride tetrahydrate
Formula: FeCl₂·4H₂O
Melting Pt: 105-110 °C
Density: 1.93
MDL Number: MFCD00149709
CAS Number: 13478-10-9
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR