Methyl 3-amino-5-fluoro-4-hydroxybenzoate ≥95%

Supplier: APOLLO SCIENTIFIC

PC502069-1G PC502069-250MG PC502069-5G
76605-958EA 578.6 USD
76605-958 76607-182 76604-498
Methyl 3-amino-5-fluoro-4-hydroxybenzoate ≥95%
Methyl 3-amino-5-fluoro-4-hydroxybenzoate
Formula: C₈H₈FNO₃
MDL Number: MFCD20725712
CAS Number: 946121-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR