Tributyrin (glycerol tributyrate) ≥97%

Supplier: APOLLO SCIENTIFIC

OR62132-100G OR62132-25G OR62132-500G
76594-504EA 78.44 USD
76594-504 76594-502 76594-506
Tributyrin (glycerol tributyrate) ≥97%
Tributyrin (Glycerol tributyrate)
Formula: C₁₅H₂₆O₆
Boiling Pt: 129…131 °C/0.03mm
Melting Pt: –75 °C
Density: 1.032
Flash Pt: 174 °C
MDL Number: MFCD00009392
CAS Number: 60-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR