1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride ≥95%

Supplier: APOLLO SCIENTIFIC
Warning

OR30668-1G OR30668-250MG
76561-136EA 401.36 USD
76561-136 76560-684
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride ≥95%
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride
Formula: C₈H₁₄ClNO₂
MDL Number: MFCD11869757
CAS Number: 374794-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR