Water, GemPure™ Select USP, purified, sterile
Supplier: GeminiBio
Synonyms:
Dihydrogen oxide
901-005-1X6
901-005-021
901-005-203
76683-188CS
103.92
USD
76683-188
76683-190
76683-192
Water, GemPure™ Select USP, purified, sterile
Water
GemPure™ Select Purified Water, Sterile (USP Grade) is manufactured and tested in accordance with the USP monograph for sterile purified water. Purified water is suitable in various non-parenteral applications, non-active medical device manufacture and cleaning applications.
- Manufactured in an ISO 13485:2016 and 21CFR820 compliant facility
- Available in 1, 20, 200 L, as well as custom sizes to suit your needs
- For further manufacturing or research use only
- Not for direct therapeutic use
Meets the USP monograph for sterile purfied water.
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 UN: 2201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...