Iron Clad Electromagnet
Supplier: EISCO SCIENTIFIC LLC
HPEMGS
470345-048EA 59.75 USD
470345-048
Iron Clad Electromagnet
Educational Materials Physics Educational Materials Magnetic Materials
Small-sized iron-clad electromagnet.
  • Lifts up to 80 lbs.
  • Powered by one (1) D cell battery
  • Comes with large hooks
  • Batteries not included
This battery powered electromagnet means serious business. Powered by (1) 'D' battery. It comes with large hooks on either end of the apparatus. Lifts up to 80 lbs. Batteries not included.