10-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-10H-phenothiazine 97%

Supplier: Ambeed
Warning

A1373476-100MG A1373476-1G A1373476-250MG A1373476-5G
76682-438EA 81 USD
76682-438 76682-440 76682-442 76682-444
10-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-10H-phenothiazine 97%
10-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-10H-phenothiazine

Caution: for Research Use Only

Formula: C₂₄H₂₄BNO₂S
CAS Number: 1771775-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR