2-Cyclopentyl-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1338826-250MG A1338826-5G A1338826-1G
76681-982EA 7 USD
76681-982 76681-984 77867-362
2-Cyclopentyl-1,3,2-dioxaborinane 98%
2-Cyclopentyl-1,3,2-dioxaborinane

Caution: for Research Use Only

Formula: C₈H₁₅BO₂
CAS Number: 30169-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR