Tri-tert-butyl 2,2',2''-(10-(2-((2-aminoethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 98%

Supplier: Ambeed
Warning

A1257133-100MG A1257133-250MG A1257133-25MG A1257133-50MG
76680-446EA 107 USD
76680-446 76680-448 76680-450 76680-452
Tri-tert-butyl 2,2',2''-(10-(2-((2-aminoethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 98%
Tri-tert-butyl 2,2',2''-(10-(2-((2-aminoethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

Caution: for Research Use Only

Formula: C₃₀H₅₈N₆O₇
MDL Number: MFCD30533300
CAS Number: 173308-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR