Tris(dibenzylideneacetone)dipalladium(0) 98% + (Carbon 64.6-69.2%)
Supplier: Ambeed
|
Danger
|
Synonyms:
Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]
A1233015-100MG
A1233015-250MG
76679-820EA
8
USD
76679-820
76679-822
Tris(dibenzylideneacetone)dipalladium(0) 98% + (Carbon 64.6-69.2%)
Tris(dibenzylideneacetone)dipalladium(0)
Caution: for Research Use Only
Formula:
C₅₁H₄₂O₃Pd₂ MW: 915.72 g/mol |
MDL Number:
MFCD00013310 CAS Number: 51364-51-3 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...