8-Bromoguanosine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

A121511-1G A121511-5G A121511-25G
76678-210EA 7 USD
76678-210 76678-212 77620-932
8-Bromoguanosine 97%
8-Bromoguanosine

Caution: for Research Use Only

Formula: C₁₀H₁₂BrN₅O₅
MW: 362.14 g/mol
MDL Number: MFCD00037985
CAS Number: 4016-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR