Aluminium distearate 4-6% Al

Supplier: Ambeed
Warning

A1159067-1000G A1159067-100G A1159067-500G
76675-464EA 23 USD
76675-464 76675-466 76675-468
Aluminium distearate 4-6% Al
Aluminium distearate

Caution: for Research Use Only

Formula: C₃₆H₇₁AlO₅
MDL Number: MFCD00055007
CAS Number: 300-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR