Potassium lactate 60% in water

Supplier: Ambeed
Warning

A1150428-1000G A1150428-100G A1150428-500G
76674-276EA 53 USD
76674-276 76674-278 76674-280
Potassium lactate 60% in water
Potassium lactate

Caution: for Research Use Only

Formula: C₃H₅O₃K
MDL Number: MFCD00036410
CAS Number: 996-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR