Polyethylene glycol monomethyl ether, M.W. ∼550

Supplier: Ambeed
Warning

Synonyms: Methoxypolyethylene glycols-average M.W. 1000mPEGMethoxy poly(ethylene glycol)Methoxypolyethylene glycolsm-PEG-OHPEG MMEPoly(ethylene glycol) methyl ether

A1146968-500G A1146968-1000G A1146968-100G A1146968-25G
76673-490EA 23.34 USD
76673-490 76673-484 76673-488 76673-486
Polyethylene glycol monomethyl ether, M.W. ∼550
Polyethylene glycol monomethyl ether

Caution: for Research Use Only

Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR