Compound E 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-(3,5-Difluorophenyl)acetamido)-N-((S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanamide

A1145654-5MG A1145654-1MG
76673-298EA 150.65 USD
76673-298 76673-296
Compound E 98%
Compound E

Caution: for Research Use Only

Formula: C₂₇H₂₄F₂N₄O₃
MW: 490.511 g/mol
MDL Number: MFCD04974531
CAS Number: 209986-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR