tert-Butyl 5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 97%

Supplier: Ambeed
Warning

A1670899-1G A1670899-250MG A1670899-5G
76669-332EA 145 USD
76669-332 76669-334 76669-336
tert-Butyl 5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 97%
tert-Butyl 5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Caution: for Research Use Only

Formula: C₁₈H₂₄BClN₂O₄
MDL Number: MFCD34563941
CAS Number: 2882875-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR