3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 95%

Supplier: Ambeed
Warning

Synonyms: Thiazolyl blue tetrazolium bromide2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromideMTT

A1518164-25G
76667-590EA 185.66 USD
76667-590
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 95%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

Caution: for Research Use Only

Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
MDL Number: MFCD00011964
CAS Number: 298-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR