Tri-tert-butyl 2,2',2''-(10-(2-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 95%

Supplier: Ambeed
Warning

A1499175-100MG A1499175-1G A1499175-250MG A1499175-50MG
76666-840EA 166 USD
76666-840 76666-842 76666-844 76666-846
Tri-tert-butyl 2,2',2''-(10-(2-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 95%
Tri-tert-butyl 2,2',2''-(10-(2-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

Caution: for Research Use Only

Formula: C₃₄H₅₈N₆O₉
CAS Number: 1613382-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR