Tri-tert-butyl 2,2',2''-(10-(2-(but-3-yn-1-ylamino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 98%

Supplier: Ambeed
Warning

A1499150-100MG A1499150-250MG A1499150-50MG
76666-826EA 246 USD
76666-826 76666-828 76666-830
Tri-tert-butyl 2,2',2''-(10-(2-(but-3-yn-1-ylamino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate 98%
Tri-tert-butyl 2,2',2''-(10-(2-(but-3-yn-1-ylamino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

Caution: for Research Use Only

Formula: C₃₂H₅₇N₅O₇
CAS Number: 2125661-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR