5-Amino-2,4,6-triiodoisophthalic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2,4,6-triiodoisophthalic Acid

A1486201-25G A1486201-5G
76665-064EA 18 USD
76665-064 76665-066
5-Amino-2,4,6-triiodoisophthalic acid 98+%
5-Amino-2,4,6-triiodoisophthalic acid

Caution: for Research Use Only

Formula: C₈H₄I₃NO₄
MDL Number: MFCD00190167
CAS Number: 35453-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR