4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)methyl)-7H-pyrrolo[2,3-d]pyrimidine 99%
Supplier: Ambeed
|
Warning
|
A1483733-100G
A1483733-1G
A1483733-250MG
A1483733-25G
A1483733-5G
76664-536EA
1187
USD
76664-536
76664-538
76664-540
76664-542
76664-544
4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)methyl)-7H-pyrrolo[2,3-d]pyrimidine 99%
4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)methyl)-7H-pyrrolo[2,3-d]pyrimidine
Caution: for Research Use Only
Formula:
C₁₅H₂₁N₅OSi |
MDL Number:
MFCD11857754 CAS Number: 941685-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...