2-Amino-4-methylpentanamide hydrochloride 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Leu-NH₂·HCl
, H-DL-Leu-NH2.HCl
A1482480-1G
A1482480-100G
A1482480-25G
A1482480-5G
76664-330EA
66.95
USD
76664-330
76664-326
76664-324
76664-328
2-Amino-4-methylpentanamide hydrochloride 98+%
2-Amino-4-methylpentanamide hydrochloride
Caution: for Research Use Only
Formula:
C₆H₁₅ClN₂O MW: 166.649 g/mol |
MDL Number:
MFCD00070244 CAS Number: 10466-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...