2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 98+%
Supplier: Ambeed
|
Warning
|
A1481692-100G
A1481692-25G
A1481692-5G
76664-230EA
175
USD
76664-230
76664-232
76664-234
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 98+%
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Caution: for Research Use Only
Formula:
C₁₁H₁₀ClN₃OS MW: 267.73 g/mol |
MDL Number:
MFCD10000630 CAS Number: 302964-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...