2'-Bromo-6'-fluoroacetophenone 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromo-6-fluorophenyl)ethanone

A1477220-100G A1477220-1G A1477220-25G A1477220-5G
76663-958EA 795 USD
76663-958 76663-960 76663-962 76663-964
2'-Bromo-6'-fluoroacetophenone 98+%
2'-Bromo-6'-fluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD13193629
CAS Number: 928715-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR