2-(2-Aminothiazol-4-yl)acetic acid 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Amino-4-thiazolyl)acetic acid, 2-Amino-4-thiazoleacetic acid
A1476017-1000G
A1476017-100G
A1476017-25G
A1476017-500G
76663-562EA
112
USD
76663-562
76663-564
76663-566
76663-568
2-(2-Aminothiazol-4-yl)acetic acid 99%
2-(2-Aminothiazol-4-yl)acetic acid
Caution: for Research Use Only
Formula:
C₅H₆N₂O₂S MW: 158.18 g/mol |
MDL Number:
MFCD00010068 CAS Number: 29676-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...