Iron(III) meso-tetraphenylporphine chloride 96%

Supplier: Ambeed
Warning

Synonyms: Iron(III) Tetraphenylporphyrin chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride, meso-Tetraphenylporphyrin iron(III) chloride, iron(III) chloride TPP, 21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride

A1467965-100MG A1467965-250MG A1467965-5G A1467965-1G
76662-630EA 14.86 USD
76662-630 76662-632 76662-634 77549-800
Iron(III) meso-tetraphenylporphine chloride 96%
Iron(III) meso-tetraphenylporphine chloride

Caution: for Research Use Only

Formula: C₄₄H₂₈ClFeN₄
MW: 704.02 g/mol
MDL Number: MFCD00012152
CAS Number: 16456-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR