Sodium diphenylamine-4-sulfonate indicator
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium N-phenylsulphanilate
, Sodium Diphenylaminesulfonate
, Sodium (4-anilinobenzenesulfonate)
, Sodium (4-anilinobenzenesulphonate)
, Sodium Diphenylaminesulphonate
, Diphenylamine-4-sulphonic acid, sodium salt
, Diphenylamine-4-sulfonic acid, sodium salt
, Sodium N-phenylsulfanilate
A1458174-500G
A1458174-1000G
A1458174-25G
A1458174-100G
76661-848EA
11.67
USD
76661-848
76661-850
76661-844
76661-846
Sodium diphenylamine-4-sulfonate indicator
Sodium diphenylamine-4-sulphonate
Caution: for Research Use Only
Formula:
C₁₂H₁₀NNaO₃S MW: 271.27 g/mol |
MDL Number:
MFCD00036468 CAS Number: 6152-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...