1-Hexadecanol 99%

Supplier: Ambeed
Warning

Synonyms: Cetyl alcohol, Palmityl alcohol

A1456728-100G A1456728-25G A1456728-500G
76661-782EA 6 USD
76661-782 77566-962 77597-270
1-Hexadecanol 99%
1-Hexadecanol

Caution: For research use only.

Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
MDL Number: MFCD00004760
CAS Number: 36653-82-4
Merck Index: 13,02037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR