Oleic acid 85%

Supplier: Ambeed
Warning

Synonyms: (9Z)-9-Octadecenoic acid

A1453784-25G A1453784-500G A1453784-100G A1453784-1000G
77622-168EA 21.63 USD
77622-168 76661-620 76661-622 76661-624
Oleic acid 85%
Oleic acid

Caution: for Research Use Only

Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
MDL Number: MFCD00064242
CAS Number: 112-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR