Sodium dihydrogen phosphate monohydrate 99%+, Analytical reagent (AR)

Supplier: Ambeed
Warning

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

A1453167-100G A1453167-25G A1453167-500G A1453167-1000G
76661-550EA 8 USD
76661-550 76661-552 76661-554 77597-242
Sodium dihydrogen phosphate monohydrate 99%+, Analytical reagent (AR)
Sodium dihydrogen phosphate monohydrate

Caution: for Research Use Only

Formula: NaH₂PO₄·H₂O
MDL Number: MFCD00149208
CAS Number: 10049-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR