98+% (mixture of isomers)

Supplier: Ambeed
Warning

A1452012-25G A1452012-5G
76661-412EA 30 USD
76661-412 76661-414
98+% (mixture of isomers)
2-Ethyl-3,5-dimethylpyrazine

Caution: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD00047392
CAS Number: 13925-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR