(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99% +(99.5%ee)

Supplier: Ambeed
Warning

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

A1443762-100G A1443762-1G A1443762-25G A1443762-5G A1443762-500G
76661-210EA 231 USD
76661-210 76661-212 76661-214 77566-912 77874-866
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99% +(99.5%ee)
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Caution: For research use only.

Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR