Ponceau S BS
Supplier: Ambeed
|
Warning
|
Synonyms:
CI 27195
, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt
, Ponceau
, 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt
, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt
A1372055-25G
A1372055-500G
A1372055-100G
76659-836EA
159.14
USD
76659-836
76659-832
76659-834
Ponceau S BS
Ponceau S
Caution: for Research Use Only
Formula:
C₂₂H₁₂N₄Na₄O₁₃S₄ MW: 760.56 g/mol |
MDL Number:
MFCD00003892 CAS Number: 6226-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...