Tetrakis[(S)-[(1S)-1-(4-bromophenyl)-2,2-diphenylcyclopropanecarboxylato]dirhodium(II) 98%

Supplier: Ambeed
Warning

A1371104-50MG A1371104-100MG
76659-668EA 320 USD
76659-668 77621-882
Tetrakis[(S)-[(1S)-1-(4-bromophenyl)-2,2-diphenylcyclopropanecarboxylato]dirhodium(II) 98%
Tetrakis[(S)-[(1S)-1-(4-bromophenyl)-2,2-diphenylcyclopropanecarboxylato]dirhodium(II)

Caution: for Research Use Only

MDL Number: MFCD27978420
CAS Number: 1345974-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR