(S)-2-Isopropyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%

Supplier: Ambeed
Warning

A1371060-100MG A1371060-1G A1371060-250MG
76659-622EA 33 USD
76659-622 76659-624 76659-626
(S)-2-Isopropyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%
(S)-2-Isopropyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole

Caution: for Research Use Only

Formula: C₁₂H₁₄N₂S
CAS Number: 1214921-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR