N1,N1,N1,N6,N6,N6-Hexamethylhexane-1,6-diaminium chloride dihydrate 97%
Supplier: Ambeed
|
Warning
|
A1370954-100G
A1370954-25G
A1370954-5G
76659-614EA
308
USD
76659-614
76659-616
76659-618
N1,N1,N1,N6,N6,N6-Hexamethylhexane-1,6-diaminium chloride dihydrate 97%
N1,N1,N1,N6,N6,N6-Hexamethylhexane-1,6-diaminium chloride dihydrate
Caution: for Research Use Only
Formula:
C₁₂H₃₄Cl₂N₂O₂ |
MDL Number:
MFCD00211306 CAS Number: 16986-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...