Isochroman-3-ol 95%

Supplier: Ambeed
Warning

A1363039-100MG A1363039-1G A1363039-250MG
76659-176EA 36 USD
76659-176 76659-178 76659-180
Isochroman-3-ol 95%
Isochroman-3-ol

Caution: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD09909385
CAS Number: 42900-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR