Silicone Flask Stand
Supplier: United Scientific Supplies
UNFLSTSIL
470349-812EA 71.35 USD
470349-812
Silicone Flask Stand
Supports Flask Supports
Hold flasks of all sizes upright without tipping over.
  • Non-slip design holds flasks securely
  • 12 steps to accommodate various flask sizes
  • Holds round bottom flasks up to 2000 ml
This silicone flask stand can support round bottom flasks as large as 2000ml, as well as other items like beakers and dishes. The stand has 12 molded-in steps to provide a secure grip for a wide range of flasks. The silicone material is heat resistant to 250 °C. Flask stand has a diameter of 16 cm and is 5 cm tall.