tert-Butyl 10-bromodecanoate 97%

Supplier: Ambeed
Warning

A944295-100MG A944295-1G A944295-250MG A944295-5G
76655-912EA 12 USD
76655-912 76655-914 76655-916 77599-284
tert-Butyl 10-bromodecanoate 97%
tert-Butyl 10-bromodecanoate

Caution: for Research Use Only

Formula: C₁₄H₂₇BrO₂
MDL Number: MFCD31728409
CAS Number: 1644575-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR