Polyethylene glycol 1,500 ∼ Mn 1500

Supplier: Ambeed
Warning

A939875-1000G A939875-500G
76655-758EA 17 USD
76655-758 76655-760
Polyethylene glycol 1,500 ∼ Mn 1500
Polyethylene glycol 1,500

Caution: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR