Oleic acid 90%

Supplier: Ambeed
Warning

Synonyms: (9Z)-9-Octadecenoic acid

A781601-100G A781601-25G
77569-474EA 9.27 USD
77569-474 76653-006
Oleic acid 90%
Oleic acid

Caution: For research use only.

Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
MDL Number: MFCD00064242
CAS Number: 112-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR