Methyl 5-nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Supplier: Ambeed
|
Warning
|
A734975-250MG
A734975-5G
A734975-100MG
76652-038EA
21
USD
76652-038
77783-815
77783-814
Methyl 5-nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Methyl 5-nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Caution: for Research Use Only
Formula:
C₁₄H₁₈BNO₆ MW: 307.1068 g/mol |
MDL Number:
MFCD23379498 CAS Number: 1393477-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...