Amyl laurate 97% (by GC)
Supplier: Ambeed
|
Warning
|
Synonyms:
Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester, Amyl Laurate
A726099-100G
A726099-25G
A726099-5G
76651-882EA
233
USD
76651-882
76651-884
77617-972
Amyl laurate 97% (by GC)
Amyl laurate
Caution: for Research Use Only
Formula:
C₁₇H₃₄O₂ MW: 270.46 g/mol |
MDL Number:
MFCD00059218 CAS Number: 5350-03-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...