Methyl 4-(hydroxymethyl)bicyclo[2.2.1]heptane-1-carboxylate 98%

Supplier: Ambeed
Warning

A602274-100MG A602274-250MG A602274-1G A602274-5G
76650-076EA 47 USD
76650-076 76650-078 77783-498 77783-499
Methyl 4-(hydroxymethyl)bicyclo[2.2.1]heptane-1-carboxylate 98%
Methyl 4-(hydroxymethyl)bicyclo[2.2.1]heptane-1-carboxylate

Caution: for Research Use Only

Formula: C₁₀H₁₆O₃
MDL Number: MFCD26727798
CAS Number: 1350821-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR