Polyethylene glycol 200 ∼ Mn 200

Supplier: Ambeed
Warning

Synonyms: Ethylene glycol polymer average Mn 200PEG 200

A388053-1000G A388053-500G A388053-100G
76648-694EA 6.37 USD
76648-694 77598-466 76648-692
Polyethylene glycol 200 ∼ Mn 200
Polyethylene glycol 200

Caution: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR