Polyethylene glycol 1,000 Mn 900-1100
Supplier: Ambeed
|
Warning
|
Synonyms:
PEG 1,000
, PEG 900-1100
A273691-1000G
A273691-500G
76647-908EA
11.33
USD
76647-908
76647-910
Polyethylene glycol 1,000 Mn 900-1100
Polyethylene glycol 1,000
Caution: for Research Use Only
Formula:
H(OCH₂CH₂)nOH MW: 1000 g/mol |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...